Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NR1D1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501729499 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-9499 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-9499 Supplier Proteintech Group Inc Supplier No. 145061AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This is a rabbit polyclonal antibody raised against the N terminus of human NR1D1.

This gene encodes a member of the nuclear hormone receptor family, specifically the NR1 subfamily of receptors. The encoded protein functions as a transcriptional repressor and may play a role in circadian rhythms and carbohydrate and lipid metabolism. Alternatively spliced transcript variants have been described.

Specifications

Antigen NR1D1
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.26 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene NR1D1
Gene Accession No. P20393, Q3UV55
Gene Alias ear 1, EAR1, hRev, NR1D1, Rev erbA alpha, Reverb alpha, THRA1, THRAL, V erbA related protein 1
Gene Symbols NR1D1
Host Species Rabbit
Immunogen NR1D1 Fusion Protein Ag5964
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 217166, 9572
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.