Learn More
NR1D2 Mouse anti-Human, Clone: OTI3D4, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF806588

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a member of the nuclear hormone receptor family, specifically the NR1 subfamily of receptors. The encoded protein functions as a transcriptional repressor and may play a role in circadian rhythms and carbohydrate and lipid metabolism. Alternatively spliced transcript variants have been described.Specifications
NR1D2 | |
Monoclonal | |
Unconjugated | |
NR1D2 | |
BD73, EAR-1R, RVR | |
Mouse | |
Affinity Chromatography | |
RUO | |
9975 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI3D4 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q14995 | |
NR1D2 | |
Human recombit protein fragment corresponding to amino acids 1-294 of human NR1D2 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.