Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NR1D2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530342
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530342 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530342 Supplier Invitrogen™ Supplier No. PA530342
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1. Predicted reactivity: Human (98%), Mouse (84%), Rat (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the nuclear hormone receptor family, specifically the NR1 subfamily of receptors. The encoded protein functions as a transcriptional repressor and may play a role in circadian rhythms and carbohydrate and lipid metabolism. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NR1D2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene NR1D2
Gene Accession No. Q14995
Gene Alias BD73; EAR-1R; EAR4; HZF2; HZF-2; Nr1d2; nuclear receptor Rev-ErbA beta variant 1; nuclear receptor Rev-ErbA beta variant 2; nuclear receptor subfamily 1 group D member 2; nuclear receptor subfamily 1, group D, member 2; orphan nuclear hormone receptor BD73; orphan nuclear receptor HZF-2; orphan nuclear receptor RVR; orphan receptor; Rev-erb; Rev-erb alpha-related receptor; Rev-erb beta; Rev-erba beta; rev-erba-alpha-related receptor; Rev-erbabeta; Rev-ErbA-beta; rev-erb-beta; Rev-erb-Beta2; RVR; V-erbA-related protein 1-related
Gene Symbols NR1D2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human NR1D2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9975
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.