Learn More
Invitrogen™ NR1D2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA532948
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Mouse, Rat, Bovine, Dog, Bat, Panda, Horse, Rabbit, Turkey, Chicken (100%); Elephant, Platypus (94%); Opossum, Xenopus (81%). For IHC(P), use proteinase K antigen retrieval. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
This gene encodes a member of the nuclear hormone receptor family, specifically the NR1 subfamily of receptors. The encoded protein functions as a transcriptional repressor and may play a role in circadian rhythms and carbohydrate and lipid metabolism. Alternatively spliced transcript variants have been described.
Specifications
NR1D2 | |
Polyclonal | |
Unconjugated | |
NR1D2 | |
BD73; EAR-1R; EAR4; HZF2; HZF-2; Nr1d2; nuclear receptor Rev-ErbA beta variant 1; nuclear receptor Rev-ErbA beta variant 2; nuclear receptor subfamily 1 group D member 2; nuclear receptor subfamily 1, group D, member 2; orphan nuclear hormone receptor BD73; orphan nuclear receptor HZF-2; orphan nuclear receptor RVR; orphan receptor; Rev-erb; Rev-erb alpha-related receptor; Rev-erb beta; Rev-erba beta; rev-erba-alpha-related receptor; Rev-erbabeta; Rev-ErbA-beta; rev-erb-beta; Rev-erb-Beta2; RVR; V-erbA-related protein 1-related | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100060122, 100349421, 259241, 353187, 396125, 485643, 532076, 9975 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q14995, Q60674, Q63504 | |
NR1D2 | |
Synthetic 16 amino acid peptide from internal region of human NR1D2. | |
50 μg | |
Primary | |
Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Horse, Chicken, Bat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.