Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NR1D2 Recombinant Rabbit Monoclonal Antibody (0Z4F1)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA551339
Description
Immunogen sequence: ALPAQEQLRP KPQLEQENIK SSSPPSSDFA KEEVIGMVTR AHKDTFMYNQ EQQENSAESM QPQRGERIPK NMEQYNLNHD H.
This gene encodes a member of the nuclear hormone receptor family, specifically the NR1 subfamily of receptors. The encoded protein functions as a transcriptional repressor and may play a role in circadian rhythms and carbohydrate and lipid metabolism. Alternatively spliced transcript variants have been described.
Specifications
NR1D2 | |
Recombinant Monoclonal | |
0.54 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
Q60674, Q63504 | |
NR1D2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 220-300 of human Rev-erb beta/NR1D2 (NP_005117.3). | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
Liquid |
ELISA, Western Blot | |
0Z4F1 | |
Unconjugated | |
NR1D2 | |
BD73; EAR-1R; EAR4; HZF2; HZF-2; Nr1d2; nuclear receptor Rev-ErbA beta variant 1; nuclear receptor Rev-ErbA beta variant 2; nuclear receptor subfamily 1 group D member 2; nuclear receptor subfamily 1, group D, member 2; orphan nuclear hormone receptor BD73; orphan nuclear receptor HZF-2; orphan nuclear receptor RVR; orphan receptor; Rev-erb; Rev-erb alpha-related receptor; Rev-erb beta; Rev-erba beta; rev-erba-alpha-related receptor; Rev-erbabeta; Rev-ErbA-beta; rev-erb-beta; Rev-erb-Beta2; RVR; V-erbA-related protein 1-related | |
Rabbit | |
Affinity chromatography | |
RUO | |
259241, 353187 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction