Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NR1H2 Mouse anti-Human, Porcine, Clone: 1E12B10, Proteintech
SDP

Catalog No. 501736432 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6432 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6432 Supplier Proteintech Group Inc Supplier No. 603451IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LXR-B belongs to the Liver X Receptor family that encodes highly homologous transcription factors. Like the highly homologous LXR-A, LXR-B forms heterodimers with the retinoic acid receptor RXRalpha, which function as sensors for cellular oxysterols which when activated, increase the expression of genes that control sterol and fatty acid metabolism and homeostasis. Recent experiments have indicated that the LXRs can also modulate both innate and adaptive immune responses. Other studies suggest that genetic variability at the LXR-B gene locus may be a risk factor for Alzheimer′s disease. One hypothesis postulates that LXR may be upstream of ApoE and potentiates the risk associated effects of the epsilon3 allele.

Specifications

Antigen NR1H2
Applications Western Blot
Classification Monoclonal
Clone 1E12B10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NR1H2
Gene Accession No. P55055
Gene Alias Liver X receptor beta, LXR b, LXRB, NER, NER I, NR1H2, Nuclear receptor NER, Oxysterols receptor LXR beta, RIP15, UNR
Gene Symbols NR1H2
Host Species Mouse
Immunogen NR1H2 Fusion Protein Ag5635
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100525290, 7376
Target Species Human, Pig
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.