Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nr1h5 Polyclonal Antibody

Catalog No. PIPA568924
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568924 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568924 Supplier Invitrogen™ Supplier No. PA568924

Rabbit Polyclonal Antibody

This target displays homology in the following species: Mouse: 100%; Rat: 100%.

Nr1h5 gene ontology annotations related to this gene include cell differentiation; cellular response to lipopolysaccharide; cholesterol homeostasis; lipid homeostasis; lipid metabolic process; multicellular organism development; negative regulation of transcription by RNA polymerase II; positive regulation of transcription by RNA polymerase II.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nr1h5
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Nr1h5
Gene Accession No. 0
Gene Alias Fxrb; Nr1h5; Nr1h5p; nuclear receptor subfamily 1, group H, member 5; similar to nuclear receptor subfamily 1, group H, member 5 isoform 2
Gene Symbols Nr1h5
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of mouse Nr1h5.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 381463
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.