Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NR1I2 Mouse anti-Human, Clone:6H11D8, Abnova™

Catalog No. 89334513 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-513 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-513 Supplier Abnova Corporation Supplier No. MAB17828
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human NR1I2.

This gene product belongs to the nuclear receptor superfamily, members of which are transcription factors characterized by a ligand-binding domain and a DNA-binding domain. The encoded protein is a transcriptional regulator of the cytochrome P450 gene CYP3A4, binding to the response element of the CYP3A4 promoter as a heterodimer with the 9-cis retinoic acid receptor RXR. It is activated by a range of compounds that induce CYP3A4, including dexamethasone and rifampicin. Several alternatively spliced transcripts encoding different isoforms, some of which use non-AUG (CUG) translation initiation codon, have been described for this gene. Additional transcript variants exist, however, they have not been fully characterized. [provided by RefSeq]

Specifications

Antigen NR1I2
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 6H11D8
Conjugate Unconjugated
Description nuclear receptor subfamily 1, group I, member 2
Dilution ELISA (1:10000)
Flow Cytometry (1:200-1:400)
Immunocytochemistry
Immunohistochemistry
Western Blot (1:500-1:2000)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene NR1I2
Gene Alias BXR, ONR1, PAR, PAR1, PAR2, PARq, PRR, PXR, SAR, SXR
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1-142 of human NR1I2 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 8856
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.