Learn More
NR2C1 Mouse anti-Human, Clone: OTI1C5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF803457

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a nuclear hormone receptor characterized by a highly conserved DNA binding domain (DBD), a variable hinge region, and a carboxy-terminal ligand binding domain (LBD) that is typical for all members of the steroid/thyroid hormone receptor superfamily. This protein also belongs to a large family of ligand-inducible transcription factors that regulate gene expression by binding to specific DNA sequences within promoters of target genes. Multiple alternatively spliced transcript variants have been described, but the full-length nature of some of these variants has not been determined.Specifications
NR2C1 | |
Monoclonal | |
Unconjugated | |
NR2C1 | |
TR2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
7181 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI1C5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P13056 | |
NR2C1 | |
Human recombit protein fragment corresponding to amino acids 1-237 of human NR2C1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.