Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NR2C2 Monoclonal Antibody (OTI4B1)
GREENER_CHOICE

Catalog No. PIMA526855
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526855 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526855 Supplier Invitrogen™ Supplier No. MA526855
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NR2C2 Monoclonal Antibody for Western Blot, IHC (P)

Members of the nuclear hormone receptor family, such as NR2C2, act as ligand-activated transcription factors. The proteins have an N-terminal transactivation domain, a central DNA-binding domain with 2 zinc fingers, and a ligand-binding domain at the C terminus. The activated receptor/ligand complex is translocated to the nucleus where it binds to hormone response elements of target genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NR2C2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4B1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NR2C2
Gene Accession No. P49116
Gene Alias mKIAA4145; Mtr2r1; Nr2c2; Nuclear hormone receptor TR4; nuclear receptor subfamily 2 group C member 2; nuclear receptor subfamily 2, group C, member 2; nuclear receptor subfamily 2, group H, member 2; orphan nuclear receptor TAK1; Orphan nuclear receptor TR4; orphan receptor TR4; orphan receptor, TR4; Tak1; testicular nuclear receptor 4; testicular receptor 4; TR2R1; Tr4; TR4 orphan receptor; TR4-NS orphan receptor
Gene Symbols NR2C2
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-241 of NR2C2 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7182
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.