Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NR2C2/TAK1 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS4636R

Description
Members of the nuclear hormone receptor family, such as NR2C2, act as ligand-activated transcription factors. The proteins have an N-terminal transactivation domain, a central DNA-binding domain with 2 zinc fingers, and a ligand-binding domain at the C terminus. The activated receptor/ligand complex is translocated to the nucleus where it binds to hormone response elements of target genes.Specifications
NR2C2/TAK1 | |
Polyclonal | |
Unconjugated | |
NR2C2 | |
mKIAA4145; Mtr2r1; NR2C2; Nuclear hormone receptor TR4; Nuclear receptor subfamily 2 group C member 2; nuclear receptor subfamily 2, group C, member 2; nuclear receptor subfamily 2, group H, member 2; Orphan nuclear receptor TAK1; Orphan nuclear receptor TR4; orphan receptor TR4; orphan receptor, TR4; TAK1; testicular nuclear receptor 4; Testicular receptor 4; TR2R1; Tr4 | |
Rabbit | |
Protein A | |
RUO | |
7182 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P49116 | |
NR2C2 | |
KLH conjugated synthetic peptide derived from human NR2C2/TAK1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction