Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NR2C2AP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598015
Description
NR2C2AP Polyclonal Antibody for Western Blot, IHC (P), ELISA
May act as a repressor of NR2C2-mediated transactivation by suppressing the binding between NR2C2/TR4 and the TR4-response element in target genes.
Specifications
NR2C2AP | |
Polyclonal | |
Unconjugated | |
NR2C2AP | |
2310073E15Rik; Nr2c2ap; nuclear receptor 2C2 associated protein; nuclear receptor 2C2-associated protein; repressor for TR4 transactivation; TR4 orphan receptor associated protein TRA16; TR4 orphan receptor-associated 16 kDa protein; TRA16 | |
Rabbit | |
Protein G | |
RUO | |
126382 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q86WQ0 | |
NR2C2AP | |
Recombinant Human Nuclear receptor 2C2-associated protein (7-139AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction