Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NR2E1/Tailless Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981166
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1166 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1166 Supplier Bioss Supplier No. BS11202R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Nuclear receptor TLX, a NR2 hepatocyte NF4-like receptor, has been shown to affect anterior brain differentiation, retinal development, and vision. It has been suggested that TLX targets genes for RAR beta2 and Pax2 during retinal development. Homozygous TLX mutant mice are viable at birth but exhibit reduced forebrain development and are more aggressive than wild-type mice. Mice with deleted TLX locus exhibit cerebral hypoplasia, blindness, and extreme aggression (fierce (frc) phenotype). TLX expression has been documented in mouse brain and eye. ESTs have been isolated from normal human brain libraries.

Specifications

Antigen NR2E1/Tailless
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene NR2E1
Gene Accession No. Q9Y466
Gene Alias BOS_9650; fc30e03; fc39f12; fierce; frc; HTll; Mtl1; Mtll; NR2D1; Nr2e1; Nuclear receptor subfamily 2 group E member 1; nuclear receptor subfamily 2, group E, member 1; Nuclear receptor TLX; Protein tailless homolog; tailes-related receptor; tailless; tailless homolog; Tll; Tlx; wu:fc30e03; wu:fc39f12; XTLL; zgc:100991
Gene Symbols NR2E1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human NR2E1/Tailless.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7101
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.