Learn More
NR2E3 Mouse anti-Human, Clone: OTI3G3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF806218

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
NR2E3, a NR2 Hepatocyte NF4-Like Receptor, has been suggested to affect cone cell proliferation in eye development. It has been shown to have a role in determining photoreceptor phenotype during human retinogenesis. In RD7 mice, a deletion in NR2E3 results in hereditary retinal dysplasia and degeneration. A homozygous missense mutation in NR2E3 has been linked to retinitis pigmentosa in Crypto-Jews of Belmonte. Two alternatively spliced isoforms have been documented. NR2E3 is expressed in the retina. ESTs have been isolated from cancerous human lung, colon, and ovary, and from normal human eye.Specifications
NR2E3 | |
Monoclonal | |
Unconjugated | |
NR2E3 | |
ESCS, PNR, RNR, RP37, rd7 | |
Mouse | |
Affinity Chromatography | |
RUO | |
10002 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI3G3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9Y5X4 | |
NR2E3 | |
Full length human recombit protein of human NR2E3 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.