Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NR2F1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113859
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113859 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113859 Supplier Invitrogen™ Supplier No. PA5113859
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VLFTSDACGL SDAAHIESLQ EKSQCALEEY VRSQYPNQPS RFGKLLLRLP For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 86%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 86%; Rat: 100%; Sheep: 86%; Zebrafish: 100%.

The protein encoded by this gene is a nuclear hormone receptor and transcriptional regulator. The encoded protein acts as a homodimer and binds to 5'-AGGTCA-3' repeats. Defects in this gene are a cause of Bosch-Boonstra optic atrophy syndrome (BBOAS).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NR2F1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Nr2f1
Gene Accession No. P10589
Gene Alias avian erythroblastic leukemia viral (v-erb-a) oncogene homolog-like 3; BBOAS; BBSOAS; chicken ovalbumin upstream promoter-transcription factor I; COUP transcription factor 1; COUP transcription factor I; COUP-TF I; COUP-TF1; COUPTFA; COUP-TFI; EAR3; EAR-3; Erbal3; Nr2f1; NR2F2; Nuclear receptor subfamily 2 group F member 1; nuclear receptor subfamily 2, group F, member 1; SVP44; Tcfcoup1; TFCOUP1; transcription factor COUP 1 (chicken ovalbumin upstream promoter 1, v-erb-a homolog-like 3); V-erbA-related protein 3
Gene Symbols Nr2f1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C terminal region of human NR2F1 (aa 326-375).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7025
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.