Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NR3A Mouse anti-Mouse,Rat, Unconjugated, Clone: N416/40, MilliporeSigma™

Catalog No. MABN18651MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN18651MI 100 μg
MABN18652MI 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN18651MI Supplier MilliporeSigma Supplier No. MABN1865100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NR3A Monoclonal antibody specifically detects NR3A in Mouse,Rat samples. It is validated for Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NR3A
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone N416/40
Conjugate Unconjugated
Gene Accession No. NP_001263284
Gene Alias NR3A;NMDAR3A;N-methyl-D-aspartate receptor subtype 3A
Gene Symbols Grin3a
Host Species Mouse
Immunogen Linear peptide from the N-terminal region of mouse Glutamate receptor ionotropic, NMDA3A (NR3A).
Molecular Weight of Antigen 127.54 kDa calculated.
Purification Method Protein G Purified
Quantity 100 μg
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity Clone N416/40 is a mouse monoclonal antibody that detects NMDAR3A (NR3A).
Target Species Mouse, Rat
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.