Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NR3C2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584969
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584969 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584969 Supplier Invitrogen™ Supplier No. PA584969
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSSAIVGVNS GGQSFHYRIG AQGTISLSRS ARDQSFQHLS SFPPVNTLVE SWKSHGDLSS RRSDGYPVLE YIPENVSSST L Highest antigen sequence indentity to the following orthologs: Mouse - 95%, Rat - 98%.

Steroid receptors are ligand-dependent, intracellular proteins that stimulate transcription of specific genes by binding to specific DNA sequences following activation by the appropriate hormone. Mineralocorticoids are a family of steroids, secreted by the adrenal cortex, necessary for the regulation of a number of metabolic processes including electrolyte regulation. These compounds exert their effect through their interaction with the mineralocorticoid receptor (MR) and that complex's subsequent association with DNA. Given the function of mineralocorticoids, it is not surprising to find that the kidney is a primary target organ for mineralocorticoids and that this organ has been shown to contain MR. The corresponding gene for the mineralocorticoid receptor is NR3C2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NR3C2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NR3C2
Gene Accession No. P08235
Gene Alias aldosterone receptor; MCR; Mineralocorticoid receptor; Mineralocorticoid receptor (aldosterone receptor); mineralocorticoid receptor 1; mineralocorticoid receptor 2; mineralocorticoid receptor delta; Mlr; MR; NR3C2; NR3C2VIT; nuclear hormone receptor; nuclear receptor subfamily 3 group C member 2; nuclear receptor subfamily 3, group C, member 2; nuclear receptor subfamily 3, group C, member 2 variant 3
Gene Symbols NR3C2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NR3C2. Recombinant protein control fragment (Product #RP-108024).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4306
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.