Learn More
NR4A2 Mouse anti-Human, Unconjugated, Clone: 1H5B4, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | NR4A2 |
---|---|
Clone | 1H5B4 |
Concentration | 1000 μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-223-9070
![]() ![]() |
Proteintech Group Inc
668781IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The NR4A2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NR4A2. This antibody recognizes human, mouse antigen. The NR4A2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
NR4A2 | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
4929 | |
Primary |
1H5B4 | |
Western Blot | |
Unconjugated | |
Human, Mouse | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.