Learn More
NR4A3 Mouse anti-Human, Clone: OTI2E1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF804872

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
NOR-1, a NR4 nerve growth factor-like receptor, has been shown to affect neuronal development, T-cell apoptosis, and cancer development. NOR-1 binds to NBRE (NGFI-B response element) and regulates the pro-opiomelanocortin (POMC) gene. NOR-1 is induced rapidly but transiently by a variety of stimuli. Downregulation of NOR-1 and other NGFI-B family members is correlated with drug addiction-prone behavior of Lewis rats. Three human protein isoforms have been documented: NOR-1, NOR-1beta, and NOR-1 3′ variant.Specifications
NR4A3 | |
Monoclonal | |
Unconjugated | |
NR4A3 | |
CHN, CSMF, MINOR, NOR1, TEC | |
Mouse | |
Affinity Chromatography | |
RUO | |
8013 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI2E1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q92570 | |
NR4A3 | |
Human recombit protein fragment corresponding to amino acids 332-626 of human NR4A3 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.