Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NR4A3/NOR1 Antibody (OTI5C2), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273052IR
Description
NR4A3/NOR1 Monoclonal specifically detects NR4A3/NOR1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
NR4A3/NOR1 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
NR4A3 | |
Recombinant protein fragment corresponding to amino acids 332-626 of human NR4A3/NOR1 (NP_008912) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot | |
OTI5C2 | |
Western Blot | |
CHN, CSMF, MINOR, NOR1, TEC | |
Mouse | |
Affinity Purified | |
GPCR | |
8013.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction