Learn More
NR5A1 Mouse anti-Human, Clone: OTI10E2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF806154

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Steroidogenic factor-1 (SF-1) is an orphan nuclear receptor involved in organ development and differentiation of the endocrine system. SF-1 has been shown to be essential for fetal development of the hypothalamus, pituitary gland, gonads, and the adrenal gland. SF-1 has been shown to promote transcriptional activity of many steroidogenic enzymes from the cytochrome P450 family. SF-1 contains two activation function domains, designated AF1 and AF2, common to many nuclear steroid receptors which have been shown to be necessary for interaction with other nuclear receptors as well as coactivators and corepressors including steroid receptor coactivator-1 (SRC-1), glucocorticoid receptor interacting protein 1 (GRIP1), cAMP response element binding protein (CREB) binding protein (CBP) and silencing mediator of retinoid acid and thyroid hormone receptor (SMRT).Specifications
NR5A1 | |
Monoclonal | |
Unconjugated | |
NR5A1 | |
AD4BP, ELP, FTZ1, FTZF1, POF7, SF-1, SF1, SPGF8, SRXX4, SRXY3, hSF-1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
2516 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin) | |
OTI10E2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q13285 | |
NR5A1 | |
Human recombit protein fragment corresponding to amino acids 1-262 of human NR5A1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.