Learn More
NR5A2 Mouse anti-Human, Clone: OTI3H8, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF805074

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Binds to the sequence element 5′-AACGACCGACCTTGAG-3′ of the enhancer II of hepatitis B virus genes, a critical cis-element of their expression and regulation. May be responsible for the liver-specific activity of enhancer II, probably in combination with other hepatocyte transcription factors. Key regulator of cholesterol 7-alpha-hydroxylase gene (CYP7A) expression in liver. May also contribute to the regulation of pancreas-specific genes and play important roles in embryonic development.Specifications
NR5A2 | |
Monoclonal | |
Unconjugated | |
NR5A2 | |
B1F, B1F2, CPF, FTF, FTZ-F1, FTZ-F1beta, LRH-1, LRH1, hB1F-2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
2494 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI3H8 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O00482 | |
NR5A2 | |
Human recombit protein fragment corresponding to amino acids 1-303 of human NR5A2 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.