Learn More
Invitrogen™ NR5A2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589359
Description
Immunogen sequence: SIPHLILELL KCEPDEPQVQ AKIMAYLQQE QANRSKHEKL STFGLMCKMA DQTLFSIVEW ARSSIFFREL KVDDQMKLLQ NCWSELLILD HIYRQVVHGK EGSIFLVTGQ QVDYSIIASQ AGATLNNLMS HAQELVAKLR SLQFDQREFV CLKFLVLFSL DVKNLENFQL VEGVQEQVNA ALLDYTMCNY PQQTEKFGQL LLRLPEIRAI SMQAEEYLYY KHLNGDVPYN NLLIEMLHAK RA Positive Samples: Mouse liver; Cellular Location: Nucleus.
Binds to the sequence element 5'-AACGACCGACCTTGAG-3' of the enhancer II of hepatitis B virus genes, a critical cis-element of their expression and regulation. May be responsible for the liver-specific activity of enhancer II, probably in combination with other hepatocyte transcription factors. Key regulator of cholesterol 7-alpha-hydroxylase gene (CYP7A) expression in liver. May also contribute to the regulation of pancreas-specific genes and play important roles in embryonic development.
Specifications
NR5A2 | |
Polyclonal | |
Unconjugated | |
Nr5a2 | |
Alpha-1-fetoprotein transcription factor; AU020803; B1-binding factor; b1-binding factor, hepatocyte transcription factor which activates enhancer II of hepatitis B virus; B1F; B1F2; CPF; CYP7A promoter-binding factor; D1Ertd308e; fetoprotein transcription factor; fetoprotein-alpha 1 (AFP) transcription factor; Ftf; FTF/LRH-1; FTZ F1; FTZ F1beta; FTZ-F1; FTZ-F1 beta; FTZ-F1 beta2 protein; FTZ-F1beta; hB1F; hB1F 2; hB1F-2; Hepatocytic transcription factor; hepatocytic transcription factor hB1F-3; I79_021747; live; liver nuclear receptor homolog-1 variant 2; liver receptor homolog 1; liver receptor homolog-1; Lrh1; LRH-1; Nr5a2; Nuclear receptor subfamily 5 group A member 2; nuclear receptor subfamily 5, group A, member 2; OR2.0; orphan nuclear receptor; SF-2; steroidogenic factor 2; UF2-H3B | |
Rabbit | |
Affinity chromatography | |
RUO | |
2494, 26424, 60349 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
2.68 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
O00482, P45448, Q9QWM1 | |
Nr5a2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 300-541 of human NR5A2 (NP_9955821). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.