Learn More
Invitrogen™ NR5A2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110041
Description
Immunogen sequence: SIPHLILELL KCEPDEPQVQ AKIMAYLQQE QANRSKHEKL STFGLMCKMA DQTLFSIVEW ARSSIFFREL KVDDQMKLLQ NCWSELLILD HIYRQVVHGK EGSIFLVTGQ QVDYSIIASQ AGATLNNLMS HAQELVAKLR SLQFDQREFV CLKFLVLFSL DVKNLENFQL VEGVQEQVNA ALLDYTMCNY PQQTEKFGQL LLRLPEIRAI SMQAEEYLYY KHLNGDVPYN NLLIEMLHAK RA.
Binds to the sequence element 5'-AACGACCGACCTTGAG-3' of the enhancer II of hepatitis B virus genes, a critical cis-element of their expression and regulation. May be responsible for the liver-specific activity of enhancer II, probably in combination with other hepatocyte transcription factors. Key regulator of cholesterol 7-alpha-hydroxylase gene (CYP7A) expression in liver. May also contribute to the regulation of pancreas-specific genes and play important roles in embryonic development.
Specifications
NR5A2 | |
Polyclonal | |
Unconjugated | |
Nr5a2 | |
Alpha-1-fetoprotein transcription factor; AU020803; B1-binding factor; b1-binding factor, hepatocyte transcription factor which activates enhancer II of hepatitis B virus; B1F; B1F2; CPF; CYP7A promoter-binding factor; D1Ertd308e; fetoprotein transcription factor; fetoprotein-alpha 1 (AFP) transcription factor; Ftf; FTF/LRH-1; FTZ F1; FTZ F1beta; FTZ-F1; FTZ-F1 beta; FTZ-F1 beta2 protein; FTZ-F1beta; hB1F; hB1F 2; hB1F-2; Hepatocytic transcription factor; hepatocytic transcription factor hB1F-3; I79_021747; live; liver nuclear receptor homolog-1 variant 2; liver receptor homolog 1; liver receptor homolog-1; Lrh1; LRH-1; Nr5a2; Nuclear receptor subfamily 5 group A member 2; nuclear receptor subfamily 5, group A, member 2; OR2.0; orphan nuclear receptor; SF-2; steroidogenic factor 2; UF2-H3B | |
Rabbit | |
Affinity chromatography | |
RUO | |
2494, 26424, 60349 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1.81 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O00482, P45448, Q9QWM1 | |
Nr5a2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 300-541 of human NR5A2 (NP_9955821). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.