Learn More
Invitrogen™ NR5A2 Polyclonal Antibody

Description
Recommended positive controls: mouse liver, Aspc1. Predicted reactivity: Mouse (88%), Rat (88%), Zebrafish (82%), Japanese Medaka (83%), Xenopus laevis (91%), Pig (96%), Chicken (96%), Sheep (95%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | NR5A2 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | Nr5a2 |
| Gene Accession No. | O00482 |
| Gene Alias | Alpha-1-fetoprotein transcription factor; AU020803; B1-binding factor; b1-binding factor, hepatocyte transcription factor which activates enhancer II of hepatitis B virus; B1F; B1F2; CPF; CYP7A promoter-binding factor; D1Ertd308e; fetoprotein transcription factor; fetoprotein-alpha 1 (AFP) transcription factor; Ftf; FTF/LRH-1; FTZ F1; FTZ F1beta; FTZ-F1; FTZ-F1 beta; FTZ-F1 beta2 protein; FTZ-F1beta; hB1F; hB1F 2; hB1F-2; Hepatocytic transcription factor; hepatocytic transcription factor hB1F-3; I79_021747; live; liver nuclear receptor homolog-1 variant 2; liver receptor homolog 1; liver receptor homolog-1; Lrh1; LRH-1; Nr5a2; Nuclear receptor subfamily 5 group A member 2; nuclear receptor subfamily 5, group A, member 2; OR2.0; orphan nuclear receptor; SF-2; steroidogenic factor 2; UF2-H3B |
| Gene Symbols | Nr5a2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.