Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NR5A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534017
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534017 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534017 Supplier Invitrogen™ Supplier No. PA534017
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NR5A2 Polyclonal Antibody for IHC (P)

Binds to the sequence element 5'-AACGACCGACCTTGAG-3' of the enhancer II of hepatitis B virus genes, a critical cis-element of their expression and regulation. May be responsible for the liver-specific activity of enhancer II, probably in combination with other hepatocyte transcription factors. Key regulator of cholesterol 7-alpha-hydroxylase gene (CYP7A) expression in liver. May also contribute to the regulation of pancreas-specific genes and play important roles in embryonic development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NR5A2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene Nr5a2
Gene Accession No. O00482
Gene Alias Alpha-1-fetoprotein transcription factor; AU020803; B1-binding factor; b1-binding factor, hepatocyte transcription factor which activates enhancer II of hepatitis B virus; B1F; B1F2; CPF; CYP7A promoter-binding factor; D1Ertd308e; fetoprotein transcription factor; fetoprotein-alpha 1 (AFP) transcription factor; Ftf; FTF/LRH-1; FTZ F1; FTZ F1beta; FTZ-F1; FTZ-F1 beta; FTZ-F1 beta2 protein; FTZ-F1beta; hB1F; hB1F 2; hB1F-2; Hepatocytic transcription factor; hepatocytic transcription factor hB1F-3; I79_021747; live; liver nuclear receptor homolog-1 variant 2; liver receptor homolog 1; liver receptor homolog-1; Lrh1; LRH-1; Nr5a2; Nuclear receptor subfamily 5 group A member 2; nuclear receptor subfamily 5, group A, member 2; OR2.0; orphan nuclear receptor; SF-2; steroidogenic factor 2; UF2-H3B
Gene Symbols Nr5a2
Host Species Rabbit
Immunogen Synthetic 17 amino acid peptide from C-terminal to ligand-binding domain of human NR5A2.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100009700, 100523688, 100653388, 2494, 490252, 541305
Target Species Human, Canine, Monkey, Bovine, Sheep, Pig, Horse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.