Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    NRAGE/MAGED1 Antibody (353124), HRP, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB3835H
Description
NRAGE/MAGED1 Monoclonal antibody specifically detects NRAGE/MAGED1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
| NRAGE/MAGED1 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| 0.1 mL | |
| Cancer | |
| 9500 | |
| Store at 4C in the dark. | 
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 353124 | |
| Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| DLXIN-1, MAGE tumor antigen CCF, melanoma antigen family D, 1, melanoma-associated antigen D1, Neurotrophin receptor-interacting MAGE homolog, NRAGE, NRAGE MAGE-D1 antigen | |
| E. coli-derived recombinant human NRAGE, Met1-Glu778, Accession # Q9Y5V3 | |
| RUO | |
| Primary | |
| Human | |
| IgG2b | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            