Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NRAGE/MAGED1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | NRAGE/MAGED1 |
---|---|
Applications | Western Blot, Immunocytochemistry, Immunofluorescence, KnockDown |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
NRAGE/MAGED1 Polyclonal specifically detects NRAGE/MAGED1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockdown Validated.Specifications
NRAGE/MAGED1 | |
Polyclonal | |
Rabbit | |
Cancer | |
DLXIN-1, MAGE tumor antigen CCF, melanoma antigen family D, 1, melanoma-associated antigen D1, Neurotrophin receptor-interacting MAGE homolog, NRAGE, NRAGE MAGE-D1 antigen | |
MAGED1 | |
IgG | |
Affinity Purified |
Western Blot, Immunocytochemistry, Immunofluorescence, KnockDown | |
Unconjugated | |
RUO | |
Human | |
9500 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:ARPKSAFKVQNATTKGPNGVYDFSQAHNAKDVPNTQPKAAFKSQNATPKGPNAAYDFSQAATTGELAANKSEMAFKAQNATTKVGPNATYNFSQSLNANDLANSRPKTPFKAWNDTTKAPTADTQTQNVNQAKMATSQADIETDPGISEP | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title