Learn More
Invitrogen™ NRAP Monoclonal Antibody (1E9)

Description
Peptide Sequence: LRHAQKAHQL QSDVKYKSDL NLTRGVGWTP PGSYKVEMAR RAAELANARG LGLQGAYRGA EAVEAGDHQS GEVNPDATEI LHVKKKKA.
Specifications
Specifications
| Antigen | NRAP |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 1E9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | NRAP |
| Gene Accession No. | Q86VF7 |
| Gene Alias | nebulin related anchoring protein; nebulin-related anchoring protein; nebulin-related-anchoring protein; NRAP; N-RAP |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.