Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NRAP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588772
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588772 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588772 Supplier Invitrogen™ Supplier No. PA588772
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MNVQPCSRCG YGVYPAEKIS CIDQIWHKAC FHCEVCKMML SVNNFVSHQK KPYCHAHNPK NNTFTSVYHT PLNLNVRTFP EAISGIHDQE DGEQCKSVFH WDMKSKDKEG APNRQPLANE RAYWTGYGEG NAWCPGALPD PEIVRMVEAR KSLGEEYTED YEQPRGKGSF PAMITPAYQR AKKANQLASQ VEYKRGHDER ISRFSTVVDT PELLRSKAGA QLQSDVRYTE DYEQQRGKGS FPAMITPAYQ IAKRANELAS DVRYHQQYQK EMRGMAGPAI GAEGILTREC ADQYGQGYPE; Positive Samples: SH-SY5Y.

May be involved in anchoring the terminal actin filaments in the myofibril to the membrane and in transmitting tension from the myofibrils to the extracellular matrix.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NRAP
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.36 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene NRAP
Gene Accession No. Q86VF7
Gene Alias nebulin related anchoring protein; nebulin-related anchoring protein; nebulin-related-anchoring protein; NRAP; N-RAP
Gene Symbols NRAP
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human NRAP (NP_0012483921).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4892
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.