Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NRARP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588827
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588827 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588827 Supplier Invitrogen™ Supplier No. PA588827
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSQAELSTCS APQTQRIFQE AVRKGNTQEL QSLLQNMTNC EFNVNSFGPE GQTALHQSVI DGNLELVKLL VKFGADIRLA NRDGWSALHI AAFGGHQDIV LYLITKAKYA ASGR; Positive Samples: Mouse kidney.

TRUSTED_SUSTAINABILITY

Specifications

Antigen NRARP
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Nrarp
Gene Accession No. Q91ZA8
Gene Alias 2700054M22Rik; Maternal Effect Lethal family member (mel-11)-like; NOTCH-regulated ankyrin repeat protein; notch-regulated ankyrin repeat-containing protein; NRARP
Gene Symbols Nrarp
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-114 of human NRARP (NP_0010043541).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 67122
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.