Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NRAS Antibody (06), Alexa Fluor™ 594 , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP305798AF594
Description
NRAS Monoclonal antibody specifically detects NRAS in Human samples. It is validated for Western Blot, ELISASpecifications
NRAS | |
Monoclonal | |
Alexa Fluor 594 | |
ALPS4, GTPase NRas, HRAS1, neuroblastoma RAS viral (v-ras) oncogene homolog, N-ras, N-ras protein part 4, NRAS1, NS6, Transforming protein N-Ras, v-ras neuroblastoma RAS viral oncogene homolog | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human NRAS (P01111; Met1-Cys186). | |
0.1 mL | |
Breast Cancer, Cancer, Cell Cycle and Replication, mTOR Pathway | |
4893 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
06 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction