Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NRBF2 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP100109AF405
Description
NRBF2 Polyclonal antibody specifically detects NRBF2 in Human, Mouse samples. It is validated for Western BlotSpecifications
| NRBF2 | |
| Polyclonal | |
| Western Blot | |
| Comodulator of PPAR and RXR, COPR, COPR1, COPR2, DKFZp564C1664, FLJ30395, NRBF-2, nuclear receptor binding factor 2, nuclear receptor binding factor-2, nuclear receptor-binding factor 2 | |
| The immunogen recognized by this antibody maps to a region between residue 237 and 287 of human nuclear receptor binding factor 2 using the numbering given in entry NP_110386.2 (GeneID 29982). | |
| 0.1 mL | |
| Signal Transduction | |
| 29982 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction