Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NRBF2 Monoclonal Antibody (OTI1B2)
GREENER_CHOICE

Catalog No. PIMA526631
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526631 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526631 Supplier Invitrogen™ Supplier No. MA526631
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NRBF2 Monoclonal Antibody for Western Blot, IHC (P)

NRBF2 (nuclear receptor-binding factor 2) modulates transcriptional activation by target nuclear receptors. It is involved in starvation-induced autophagy probably by its association with PI3K complex I (PI3KC3-C1). NRBF2 stabilizes the PI3KC3-C1 assembly and enhances ATG14-linked lipid kinase activity of PIK3C3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NRBF2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1B2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Nrbf2
Gene Accession No. Q96F24
Gene Alias Comodulator of PPAR and RXR; comodulator of PPAR and RXR 1; comodulator of PPAR and RXR 2; COPR; COPR1; COPR2; Nrbf2; NRBF-2; nuclear receptor binding factor 2; nuclear receptor binding factor-2; nuclear receptor-binding factor 2
Gene Symbols Nrbf2
Host Species Mouse
Immunogen Full length human recombinant protein of NRBF2 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29982
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.