Learn More
NRBF2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301851A
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predicted to react with Orangutan
May modulate transcriptional activation by target nuclear receptors. Can act as transcriptional activator (in vitro).Specifications
| NRBF2 | |
| Polyclonal | |
| Unconjugated | |
| NRBF2 | |
| COPR1, COPR2, NRBF-2, comodulator of PPAR and RXR, nuclear receptor binding factor-2, nuclear receptor-binding factor 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 29982 | |
| 4° C | |
| Liquid |
| Immunohistochemistry, Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q96F24 | |
| NRBF2 | |
| Between 125 and 175 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.