Learn More
NRBF2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Supplier:  Proteintech Group Inc 248581AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-173-4579
Only null left
Add to Cart
Description
Description
May modulate transcriptional activation by target nuclear receptors. Can act as transcriptional activator (in vitro).
Specifications
Specifications
NRBF2
Polyclonal
Unconjugated
NRBF2
Comodulator of PPAR and RXR, COPR, COPR1, COPR2, NRBF 2, NRBF2
Rabbit
Antigen Affinity Chromatography
RUO
29982, 641340
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.74 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8VCQ3, Q96F24
Nrbf2
NRBF2 Fusion Protein Ag21403
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.