Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NRBP Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556182
Description
Immunogen sequence: STTSAASPEE EEESEDESEI LEESPCGRWQ KRREEVNQRN VPGIDSAYLA MDTEEGVEVV WNEVQFSERK NYKLQEEKVR AVFDNLIQLE HLNIVKFHKY WADIKENKAR VIFITEYMSS GSLKQFLKKT Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
May play a role in subcellular trafficking between the endoplasmic reticulum and Golgi apparatus through interactions with the Rho-type GTPases. Binding to the NS3 protein of dengue virus type 2 appears to subvert this activity into the alteration of the intracellular membrane structure associated with flaviviral replication. [UniProt].
Specifications
NRBP | |
Polyclonal | |
Unconjugated | |
NRBP1 | |
B230344L17Rik; BCON3; HLS7-interacting protein kinase; Madm; MLF1 adapter molecule; MUDPNP; multiple domain putative nuclear protein; myeloid leukemia factor 1 adaptor molecule; Nrbp; NRBP1; nuclear receptor binding protein 1; Nuclear receptor-binding protein; QtsA-11263; unnamed protein product | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29959 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UHY1 | |
NRBP1 | |
Recombinant protein corresponding to Human NRBP. Recombinant protein control fragment (Product #RP-91411). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction