Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NRBP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587790
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587790 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587790 Supplier Invitrogen™ Supplier No. PA587790
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AFGLPRPQQP QQEEVTSPVV PPSVKTPTPE PAEVETRKVV LMQCNIESVE EGVKHHLTLL LKLEDKLNRH LSCDLMPNEN IPELAAELVQ LGFISEADQS RLTSLLEETL NKFNFARNST LNSAAVTVSS; Positive Samples: Jurkat, MCF-7, Mouse brain; Cellular Location: Cell projection, Cytoplasm, Endomembrane system, cell cortex, lamellipodium.

May play a role in subcellular trafficking between the endoplasmic reticulum and Golgi apparatus through interactions with the Rho-type GTPases. Binding to the NS3 protein of dengue virus type 2 appears to subvert this activity into the alteration of the intracellular membrane structure associated with flaviviral replication. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen NRBP
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.65 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NRBP1
Gene Accession No. Q99J45, Q9UHY1
Gene Alias B230344L17Rik; BCON3; HLS7-interacting protein kinase; Madm; MLF1 adapter molecule; MUDPNP; multiple domain putative nuclear protein; myeloid leukemia factor 1 adaptor molecule; Nrbp; NRBP1; nuclear receptor binding protein 1; Nuclear receptor-binding protein; QtsA-11263; unnamed protein product
Gene Symbols NRBP1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 406-535 of human NRBP1 (NP_0375241).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192292, 29959
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.