Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NRBP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598977
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598977 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598977 Supplier Invitrogen™ Supplier No. PA598977
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NRBP Polyclonal Antibody for Western Blot, IHC (P), ELISA

May play a role in subcellular trafficking between the endoplasmic reticulum and Golgi apparatus through interactions with the Rho-type GTPases. Binding to the NS3 protein of dengue virus type 2 appears to subvert this activity into the alteration of the intracellular membrane structure associated with flaviviral replication. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen NRBP
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NRBP1
Gene Accession No. Q9UHY1
Gene Alias B230344L17Rik; BCON3; HLS7-interacting protein kinase; Madm; MLF1 adapter molecule; MUDPNP; multiple domain putative nuclear protein; myeloid leukemia factor 1 adaptor molecule; Nrbp; NRBP1; nuclear receptor binding protein 1; Nuclear receptor-binding protein; QtsA-11263; unnamed protein product
Gene Symbols NRBP1
Host Species Rabbit
Immunogen Recombinant Human Nuclear receptor-binding protein (406-535AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29959
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.