Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NRBP1 Antibody (OTI5C3), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273056C
Description
NRBP1 Monoclonal specifically detects NRBP1 in Human, Mouse, Rat, Monkey, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
NRBP1 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
NRBP1 | |
Full-length protein expressed in 293T cell transfected with human NRBP1 expression vector. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Monkey, Primate | |
IgG2b |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI5C3 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
BCON3NRBPMADM, FLJ27109, FLJ35541, MUDPNP, multiple domain putative nuclear protein, myeloid leukemia factor 1 adaptor molecule, nuclear receptor binding protein, nuclear receptor binding protein 1, nuclear receptor-binding protein | |
Mouse | |
Affinity Purified | |
Protein Kinase | |
29959.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction