Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NRBP2 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 4F10H7, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | NRBP2 |
---|---|
Clone | 4F10H7 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-7104
![]() ![]() |
Proteintech Group Inc
666871IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Nrbp2 may regulate apoptosis of neural progenitor cells during their differentiation.Specifications
NRBP2 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
DKFZp434P086, NRBP2, pp9320, TRG 16, TRG16 | |
NRBP2 | |
IgG2a | |
Protein A | |
Antibody |
4F10H7 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Pig, Rat | |
Q91V36, Q9NSY0 | |
110260412, 223649, 340371, 680451 | |
NRBP2 Fusion Protein Ag17011 | |
Primary | |
-20°C | |
NRBP2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title