Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NRBP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530887
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530887 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530887 Supplier Invitrogen™ Supplier No. PA530887
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: IMR32, U87-Mg. Predicted reactivity: Mouse (93%), Rat (93%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Nrbp2 may regulate apoptosis of neural progenitor cells during their differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NRBP2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.88 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene Nrbp2
Gene Accession No. Q9NSY0
Gene Alias BC011468; NRBP2; nuclear receptor binding protein 2; nuclear receptor-binding protein 2; pp9320; Transformation-related gene 16 protein; transformation-related protein 16; TRG16; TRG-16
Gene Symbols Nrbp2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human NRBP2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340371
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.