Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NRBP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565039
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565039 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565039 Supplier Invitrogen™ Supplier No. PA565039
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MELDKFLEDVR NGIYPLMNFA ATRPLGLPRV LAPPPEEVQK AKTPTPEPFD SETRKVIQMQ CNLERSEDKA RWHLTLLLVL EDRLHRQLTY DLLPTDSAQD LASELVHYGF LHEDDRMKLA AFLESTF Highest antigen sequence identity to the following orthologs - mouse 95%, rat 95%.

Nrbp2 may regulate apoptosis of neural progenitor cells during their differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NRBP2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Nrbp2
Gene Accession No. Q9NSY0
Gene Alias BC011468; NRBP2; nuclear receptor binding protein 2; nuclear receptor-binding protein 2; pp9320; Transformation-related gene 16 protein; transformation-related protein 16; TRG16; TRG-16
Gene Symbols Nrbp2
Host Species Rabbit
Immunogen Recombinant Human NRBP2. Recombinant protein control fragment (Product #RP-104595).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340371
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.