Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NRBP2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989873
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9873 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9873 Supplier Bioss Supplier No. BS19344R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Nrbp2 may regulate apoptosis of neural progenitor cells during their differentiation.

Specifications

Antigen NRBP2
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH
Gene Nrbp2
Gene Accession No. Q9NSY0
Gene Alias BC011468; NRBP2; nuclear receptor binding protein 2; Nuclear receptor-binding protein 2; PP9320; Transformation-related gene 16 protein; transformation-related protein 16; TRG16; TRG-16
Gene Symbols Nrbp2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human NRBP2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340371
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.