Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NRBP2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA572212
Description
May regulate apoptosis of neural progenitor cells during their differentiation.
Specifications
NRBP2 | |
Polyclonal | |
Unconjugated | |
NRBP2 | |
Nuclear receptor-binding protein 2; Transformation-related gene 16 protein; TRG-16; NRBP2 (<a href=""http://www.genenames.org/cgi-bin/gene_symbol_report?hgnc_id=19339"" target=""_blank"">HGNC:19339</a>) | |
Rabbit | |
Protein A | |
RUO | |
223649, 340371 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q91V36, Q9NSY0 | |
NRBP2 | |
KLH conjugated synthetic peptide between 394-428 amino acids from the C-terminal region of human NRBP2 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction