Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NrCAM Antibody (MM0495-8E25), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP211826F
Description
NrCAM Monoclonal specifically detects NrCAM in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
NrCAM | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
NRCAM | |
Human recombinant NrCAM | |
0.1 mL | |
4897 | |
Store at 4C in the dark. | |
IgG2 |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
MM0495-8E25 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
bravo, hBravo, KIAA0343Neuronal surface protein Bravo, MGC138845, MGC138846, neuronal cell adhesion molecule, Ng-CAM-related, NgCAM-related cell adhesion molecule, nr-CAM | |
Mouse | |
Protein G purified | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction