Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NRCAM Monoclonal Antibody (N364/51), PerCP
GREENER_CHOICE

Catalog No. PIMA545683
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545683 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545683 Supplier Invitrogen™ Supplier No. MA545683
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Rat: 96% identity (795/822 amino acids identical). Human: 91% identity (753/822 amino acids identical) ~50% identity with Neurofascin. 1 μg/mL of MA5-45683 was sufficient for detection of NrCAM in 20 μg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 160kDa. This antibody was formerly sold as clone S364-51.

Neuron-glia-related cell adhesion molecule (NRCAM) is a gene located on chromosome 7q31.1, encoding a protein that belongs to the immunoglobulin superfamily of cell adhesion molecules. NRCAM is expressed in the nervous system and plays a vital role in the development and maintenance of neural connections. This adhesion molecule facilitates neuron-neuron and neuron-glia interactions, impacting processes such as axon guidance, synapse formation, and cell signaling, which are crucial for neural development and plasticity. NRCAM is involved in the modulation of signal transduction pathways that influence neuronal growth and repair, contributing to the stabilization of synaptic structures. Alterations in NRCAM expression or function have been linked to neurodevelopmental disorders, including autism spectrum disorders, and certain psychiatric conditions, reflecting its importance in maintaining neural circuit integrity. Research into NRCAM continues to explore its therapeutic potential in neurobiology and its implications in neurological and psychiatric disorders.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NRCAM
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone N364/51
Concentration 1 mg/mL
Conjugate PerCP
Formulation 2.48mM MES, 95.64mM phosphate with 2mM EDTA and no preservative; pH 7.4
Gene NRCAM
Gene Accession No. P97686, Q810U4, Q92823
Gene Alias ankyrin-binding cell adhesion molecule NrCAM; Bravo; C030017F07Rik; C130076O07Rik; hBravo; KIAA0343; mBravo; MGC138845; MGC138846; mKIAA0343; neuronal cell adhesion molecule; Neuronal surface protein Bravo; neuron-glia-CAM-related cell adhesion molecule; ng-CAM-related; NgCAM-related cell adhesion molecule; Nr CAM; Nrcam; nr-CAM; rBravo
Gene Symbols NRCAM
Host Species Mouse
Immunogen Fusion protein amino acids 30-845 (extracellular domain) of mouse NrCAM.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 319504, 4897, 497815
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.