Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nrcam Mouse anti-Human, Mouse, Rat, Biotin, Clone:S364-51, Abnova™

Catalog No. 89333315 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-315 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-315 Supplier Abnova Corporation Supplier No. MAB16877
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant mouse Nrcam.

Specifications

Antigen Nrcam
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone S364-51
Conjugate Biotin
Description neuron-glia-CAM-related cell adhesion molecule
Dilution Immunocytochemistry (1:100)
Immunofluorescence (1:100)
Western Blot (1:1000)
The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene Nrcam
Gene Alias Bravo, C030017F07Rik, C130076O07Rik, mKIAA0343
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 30-845 at extracellular domain of mouse Nrcam.
Quantity 100 μg
Gene ID (Entrez) 319504
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.