Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NrCAM Antibody (S364-51), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242191F
Description
NrCAM Monoclonal specifically detects NrCAM in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
NrCAM | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
NRCAM | |
Fusion protein amino acids 30-845 (extracellular domain) of mouse NrCAM. Rat: 96% identity (795/822 amino acids identical). Human: 91% identity (753/822 amino acids identical) ∼50% identity with Neurofascin. | |
0.1 mL | |
4897 | |
Human, Mouse, Rat | |
Purified |
Immunocytochemistry, Immunofluorescence | |
S364-51 | |
Immunocytochemistry/Immunofluorescence | |
bravo, hBravo, KIAA0343Neuronal surface protein Bravo, MGC138845, MGC138846, neuronal cell adhesion molecule, Ng-CAM-related, NgCAM-related cell adhesion molecule, nr-CAM | |
Mouse | |
Protein G purified | |
Primary | |
Approx 50% identity with NeurofascinDetects approx 160kDa. | |
Store at 4C in the dark. | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction