Learn More
NRD1/Nardilysin Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304961AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat and Orangutan
Cleaves peptide substrates on the N-terminus of arginine residues in dibasic pairs.Specifications
NRD1/Nardilysin | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
O43847, Q8BHG1 | |
Nrd1, NRDC | |
Between 275 and 325 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
NRDC | |
hNRD1, hNRD2, NRD convertase, NRD-C, nardilysin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
230598, 4898 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.